Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRMT6 Antibody (SR1719), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PRMT6 |
---|---|
Clone | SR1719 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
PRMT6 Monoclonal antibody specifically detects PRMT6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PRMT6 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.1.1.-, EC 2.1.1.125, FLJ10559, FLJ51477, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6, Histone-arginine N-methyltransferase PRMT6, HMT1 hnRNP methyltransferase-like 6, HMT1 hnRNP methyltransferase-like 6 (S. cerevisiae), HRMT1L6, protein arginine methyltransferase 6, protein arginine N-methyltransferase 6 | |
A synthesized peptide derived from human PRMT6 (Uniprot #: Q96LA8) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1719 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Chromatin Research, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
55170 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title