Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRMT6 Mouse anti-Human, Clone: PCRP-PRMT6-2C9, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen PRMT6
Clone PCRP-PRMT6-2C9
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Protein Array
Applications Flow Cytometry, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122786
View Documents
Novus Biologicals
NBP30765620UG
20 μg
Each for $138.37
Add to cart
 
NB122785
View Documents
Novus Biologicals
NBP30765610UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

PRMT6 Monoclonal antibody specifically detects PRMT6 in Human samples. It is validated for Flow Cytometry, Protein Array
Specifications

Specifications

PRMT6
0.2 mg/ml
Flow Cytometry, Peptide Array
Purified
RUO
Human
EC 2.1.1.-, EC 2.1.1.125, FLJ10559, FLJ51477, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6, Histone-arginine N-methyltransferase PRMT6, HMT1 hnRNP methyltransferase-like 6, HMT1 hnRNP methyltransferase-like 6 (S. cerevisiae), HRMT1L6, protein arginine methyltransferase 6, protein arginine N-methyltransferase 6
Recombinant full-length human PRMT6 protein
Monoclonal
Protein A or G purified
PCRP-PRMT6-2C9
Flow Cytometry 1-2 ug/million cells, Protein Array
Unconjugated
Mouse
Chromatin Research, Signal Transduction
10 mM PBS with 0.05% BSA
55170
IgG2b
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit