Learn More
PRMT6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153951AP
Catalog No. 50-173-0198
Add to cart
Description
Description
Western blot analysis using this antibody detected a major band around 42 kDa in Hela cells.
Protein arginine N-methyltransferases, such as PRMT6, catalyze the sequential transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine.
Specifications
Specifications
PRMT6
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6NZB1, Q96LA8
HRMT1L6, PRMT6
Rabbit
IgG
150 μL
-20°C
Primary
55170, 99890
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PRMT6
Liquid
PRMT6
PRMT6 Fusion Protein Ag7592
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse