Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
PRNP Rabbit anti-Human, Unconjugated, Clone: EHH-16, Abnova
Rabbit Monoclonal Antibody.
Manufacturer: Abnova Corporation MAB20620
Description
PRNP Monoclonal antibody specifically detects PRNP in Human samples. It is validated for ICC, WB-Ce, IHC-P, IF, Flow Cyt.Specifications
PRNP | |
EHH-16 | |
prion protein | |
Liquid | |
Rabbit | |
IgG | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PRNP | |
ASCR, CD230, CJD, GSS, MGC26679, PRIP, PrP, PrP27-30, PrP33-35C, PrPc, prion | |
A synthetic peptide corresponding to human PRNP | |
100 μg | |
Primary | |
5621 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title