Pro-Lab Diagnostics Microbank™ Bacterial and Fungal Preservation System

Designed to simplify the storage and retrieval of bacterial and fungal cultures.  Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.

Manufacturer: Pro-Lab Diagnostics PL.170/R

 View more versions of this product

Catalog No. 22-286-154

  Check Availability
Add to Cart

    Specifications

    Product Type Cryogenic Storage Vial
    Storage Requirements -70°C
    Closure Color Red
    For Use With (Application) Designed to simplify the storage and retrieval of bacterial and fungal cultures
    Quantity 80 Pack

    • Incorporates treated beads and a special cryopreservative solution
    • Each vial contains 25 beads, providing repeated identical cultures from the original organism
    • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
    • 80 Vials supplied in a freezer box with“see through”cover
    • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit