Pro-Lab Diagnostics Prolex™ - Blue Staph Latex Kits
A rapid, sensitive method for distinguishing Staphylococcus aureus that possess coagulase (clumping factor) and/or protein A from other species of staphylococci
Manufacturer: Pro-Lab Diagnostics PL.090
 View more versions of this product
Catalog No. 22-286-168
  Check Availability
Add to Cart
    Specifications
    Product Type Agglutination Card
    Detectable Analytes Staphylococcus aureus possessing coagulase (clumping factor), Protein A
    For Use With (Equipment) Blue Staph Latex Kit
    Quantity 50 Cards
  • Description & Specifications
Related Products
Product Content Feedback