Pro-Lab Diagnostics Vision™ Salmonella Slide Agglutination Antisera
A comprehensive range of Salmonella slide agglutination antisera
Manufacturer: Pro-Lab Diagnostics PL.6015
 View more versions of this product
Catalog No. 22-286-232
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Monovalent Somatic Group D Factor 9 Antisera Slide
    Detectable Analytes Genus Salmonella
    Shelf Life 5 years
    Storage Requirements 2° to 8°C
    Quantity 3mL
  • Description & Specifications
Related Products
Product Content Feedback