Pro-Lab Diagnostics Vision™ Polyvalent Shigella Slide Agglutination Antisera
Intended to aid in the serological identification of Shigella organisms
Manufacturer: Pro-Lab Diagnostics PL.6901
 View more versions of this product
Catalog No. 22-286-239
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Product Type Polyvalent Shigella flexneri 1 to 6 X and Y Antisera Slide
    Detectable Analytes Genus Shigella
    Quantity 2mL
    Storage Requirements 2° to 8°C
  • Description & Specifications
Related Products
Product Content Feedback