Learn More
Pro-Lab Diagnostics™ EnviroBead™ Autoclave Deodorant
Available in three pleasant fragrances
Manufacturer:  Pro-Lab Diagnostics™ PL.603/100
 View more versions of this product
Catalog No. 22-286-166
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • A convenient, easy-to-use, and economical method of neutralizing odors associated with the routine autoclaving of laboratory waste
  • Each capsule is sufficient to neutralize the odor associated with 10 cubic feet of waste
Specifications
Specifications
Peach
Yes
100 Capsules