Learn More
[Pro9] Substance P, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66198MCR
 View more versions of this product
Catalog No. AAJ66198MCR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
1387.71
1mg
Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Pro-Leu-Met-NH2
-30° to -10°C
HPLC
Solid
RUO
[Pro9] Substance P