Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proadifen hydrochloride, ≥95%, MilliporeSigma™ Supelco™

$93.00 - $336.50

Specifications

Percent Purity ≥95%
Grade Analytical Standard
Linear Formula C23H31 NO2 · HCl
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4935
View Documents
MilliporeSigma Supelco
P1061100MG
100 mg
Each for $93.00
Add to cart
 
11-101-4936
View Documents
MilliporeSigma Supelco
P1061500MG
500 mg
Each for $336.50
Add to cart
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

≥95%
Analytical Standard
C23H31 NO2 · HCl
Neat
Safety and Handling

Safety and Handling

RTECSNumber : YV7175000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit