Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proBDNF Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific OSB00015W

Catalog No. PIOSB00015W

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute with 100 μL of sterile water.

plays a role in the development of hippocampal long term potentiation; involved in regulation of synaptic plasticity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

proBDNF
Polyclonal
Bdnf
Brain-derived neurotrophic factor precursor, proBDNF, pro BDNF
Rabbit
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
P21237, P23363
BDNF
A synthetic peptide from mouse proBDNF
RUO
12064, 24225
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit