Learn More
Probenecid, 98%, Thermo Scientific Chemicals
Description
It is a uricosuric drug, that is, it increases uric acid excretion in the urine. It is primarily used in treating gout and hyperuricemia. Probenecid was developed as an alternative to caronamide to competitively inhibit renal excretion of some drugs, thereby increasing their plasma concentration and prolonging their effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water (partly miscible), solutions of alkali hydroxides,, ethanol (100 mM), acetone, chloroform, sodium hydroxide (1 M), DMSO (100 mM), methanol, dilute acids (practically insoluble), dilute alkalies, and ether (slightly soluble).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 198°C to 199°C |
Assay Percent Range | 0.98 |
Quantity | 2.5 g |
Merck Index | 14,7754 |
Solubility Information | Soluble in water (partly miscible),solutions of alkali hydroxides,,ethanol (100 mM),acetone,chloroform,sodium hydroxide (1 M),DMSO (100 mM),methanol,dilute acids (practically insoluble),dilute alkalies,and ether (slightly soluble). |
Formula Weight | 285.36 |
Percent Purity | 98% |
Chemical Name or Material | Probenecid |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 200-344-3 |
RTECSNumber | DG9400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.