Learn More
Invitrogen™ ProBond™ Nickel-Chelating Resin Available on GSA/VA Contract for Federal Government customers only.
ProBond™ Nickel-Chelating Resin
$688.00
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
R80101 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
R80101
N/A 50mL
Each for $688.00
Add to cart
Description
Description
ProBond™ Nickel-Chelating Resin is a nickel-charged affinity resin used to purify recombinant proteins containing a polyhistidine (6xHis) sequence. Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine. One-step purification can be performed under both native and denaturing conditions. ProBond™ Resin uses the chelating ligand iminodiacetic acid (IDA) coupled to a highly cross-linked 6% agarose resin that is suitable for use in FPLC, batch, and gravity flow applications.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Safety and Handling
Safety and Handling
SDS
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.