Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Probucol, Tocris Bioscience™

Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
23288-49-5
C31H48O2S2
516.843
FYPMFJGVHOHGLL-UHFFFAOYSA-N
probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum
4912
CHEBI:8427
2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C
This item is not returnable. View return policy
23288-49-5
C31H48O2S2
516.843
FYPMFJGVHOHGLL-UHFFFAOYSA-N
probucol, lorelco, biphenabid, bisphenabid, lurselle, bisbid, lesterol, lursell, panavir, probucolum
4912
CHEBI:8427
2,6-ditert-butyl-4-[2-(3,5-ditert-butyl-4-hydroxyphenyl)sulfanylpropan-2-ylsulfanyl]phenol
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)SC(C)(C)SC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C

ABCA1 (CERP) inhibitor; also antioxidant, anti-inflammatory and hypocholesterolemic agent

ABCA1 (CERP) inhibitor. Inhibits cholesterol efflux from J774 macrophages expressing ABCA1 by up to 80%. Inhibits atherogenesis in genetically hypercholesterolemic rabbits (Watanabe) and attenuates ischemia/reperfusion-induced cardiomyocyte apoptosis. Antioxidant, anti-inflammatory and hypocholesterolemic agent.
Quantity 100 mg
Formula Weight 516.84
Percent Purity >99%
Chemical Name or Material Probucol

Recommended Storage : Desiccate at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit