Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procainamide hydrochloride, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
614-39-1
C13H21N3O·HCl
271.78
MFCD00012998
ABTXGJFUQRCPNH-UHFFFAOYSA-N
procainamide hydrochloride,procainamide hcl,pronestyl,procanbid,procapan,procan,procainhydrochlorid,procan sr,procainii chloridum,promide hydrochloride
66068
CHEBI:8429
4-amino-N-[2-(diethylamino)ethyl]benzamide;hydrochloride
CCN(CC)CCNC(=O)C1=CC=C(C=C1)N.Cl
614-39-1
C13H21N3O·HCl
271.78
MFCD00012998
ABTXGJFUQRCPNH-UHFFFAOYSA-N
procainamide hydrochloride,procainamide hcl,pronestyl,procanbid,procapan,procan,procainhydrochlorid,procan sr,procainii chloridum,promide hydrochloride
66068
CHEBI:8429
4-amino-N-[2-(diethylamino)ethyl]benzamide;hydrochloride
CCN(CC)CCNC(=O)C1=CC=C(C=C1)N.Cl
Quantity 100g
Color White to Yellow
Packaging Glass bottle
Melting Point 166°C to 170°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula H2NC6H4CONHCH2CH2N(C2H5)2·HCl
Beilstein 14, III, 1077
Merck Index 15, 7874
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in chloroform,very sparingly soluble in benzene and ether
Formula Weight 271.78
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Procainamide hydrochloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-381-7