Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procainamide Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
614-39-1
C13H22ClN3O
271.79
ABTXGJFUQRCPNH-UHFFFAOYSA-N
hydrogen 4-amino-N-[2-(diethylamino)ethyl]benzamide chloride
[H+].[Cl-].CCN(CC)CCNC(=O)C1=CC=C(N)C=C1
614-39-1
C13H22ClN3O
271.79
ABTXGJFUQRCPNH-UHFFFAOYSA-N
hydrogen 4-amino-N-[2-(diethylamino)ethyl]benzamide chloride
[H+].[Cl-].CCN(CC)CCNC(=O)C1=CC=C(N)C=C1

P1729, 614-39-1, C13H21N3O•HCl

Spectrum™ Procainamide Hydrochloride, USP is a sodium channel blocker used for the medical treatment of cardiac arrhythmias. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.003
Ignition Residue 0.001
Quantity 25 g
Melting Range 165-169°C
CAS Max % 1
Percent Purity 98 to 102%
Grade USP