Learn More
Medchemexpress LLC Procaine hydrochloride | 51-05-8 | MFCD00013000 | 97.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0546A1G
Encompass_Preferred
Procaine hydrochloride is a DNA-demethylating agent that acts through multiple targets. It can reduce 5-methylcytosine DNA content by 40% and demethylate densely hypermethylated CpG islands. This agent also shows growth-inhibitory effects in cancer cells, inducing mitotic arrest, and inhibits 5-HT3 receptor-mediated inward current. It is intended for research use only.
  • DNA-demethylating agent
  • Acts through multiple targets
  • Slow onset and short duration of action
  • Reduces 5-methylcytosine DNA content by 40%
  • Demethylates densely hypermethylated CpG islands
  • Exhibits growth-inhibitory effects in cancer cells
  • Causes mitotic arrest
  • Inhibits 5-HT3 receptor-mediated inward current
  • Functions as an excitant of limbic system cells
  • Alters synaptic transmission in amygdala output pathways
  • For research use only
Catalog No. 50-004-08591