Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procaine Hydrochloride, Crystal, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
51-05-8
C13H21ClN2O2
272.77
HCBIBCJNVBAKAB-UHFFFAOYSA-N
hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1
51-05-8
C13H21ClN2O2
272.77
HCBIBCJNVBAKAB-UHFFFAOYSA-N
hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

PR110, 51-05-8, C13H20N2O2•HCl

Spectrum™ Procaine Hydrochloride, Crystal, USP is considered an anesthetic drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Bottle
Ignition Residue 0.0015
Loss on Drying 0.01
Melting Range 153-158°C
Quantity 100 g
Acidity To pass test
CAS Max % 1
Percent Purity 99 to 101%
Grade USP