Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Procalcitonin Mouse anti-Human, Clone: 14A2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP3005230.2MG
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA.Specifications
Procalcitonin | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
14A2 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
Recombinant fragment (Human). a.a.r. 72-81 of procalcitonin (PCT) | |
0.2 mg | |
Signal Transduction | |
796 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction