Learn More
Procalcitonin Mouse anti-Human, DyLight 550, Clone: 38F11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP179065R
Catalog No. NBP179065R
Add to cart
Description
Description
Procalcitonin Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications
Specifications
Procalcitonin
38F11
Western Blot, ELISA
Purified
CALCA
Human PCT
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human PCT
ELISA, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
ATP2B2plasma membrane calcium ATPase, ATPase, Ca++ transporting, plasma membrane 4, DKFZp686G08106, DKFZp686M088, EC 3.6.3, EC 3.6.3.8, Matrix-remodeling-associated protein 1, matrix-remodelling associated 1, MXRA1, Plasma membrane calcium ATPase isoform 4, plasma membrane calcium pump, Plasma membrane calcium pump isoform 4, plasma membrane calcium-transporting ATPase 4, PMCA4b, PMCA4PMCA4x, sarcolemmal calcium pump
Mouse
IgG1
0.2 ml
Cancer
Primary
796
Human