Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procalcitonin Mouse anti-Human, Rat, Alexa Fluor™ 532, Clone: 4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221620AF532

Catalog No. NP221620A32


Add to cart

Description

Description

Procalcitonin Monoclonal antibody specifically detects Procalcitonin in Human, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Procalcitonin
4
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
CALCA
Synthetic human alpha-CGRP adsorbed onto aluminum hydroxide gel. The epitope lies within residues 19-37 of human alpha-CGRP1.
Protein A or G purified
Cancer, Cellular Markers, Neuroscience, Signal Transduction
Primary
796.0
Human, Rat
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody binds human alpha-CGRP. Based on inhibition ELISA, this antibody cross-reacts up to 12% with human beta-CGRP and human amylin, while there is little (<4%) cross-reaction with rat alpha-CGRP.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit