Learn More
Procalcitonin Mouse anti-Human, Rat, Alexa Fluor™ 594, Clone: 4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP221620AF594
Catalog No. NP221620A94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Procalcitonin Monoclonal antibody specifically detects Procalcitonin in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Procalcitonin
4
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
This antibody binds human alpha-CGRP. Based on inhibition ELISA, this antibody cross-reacts up to 12% with human beta-CGRP and human amylin, while there is little (<4%) cross-reaction with rat alpha-CGRP.
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CALCA
Synthetic human alpha-CGRP adsorbed onto aluminum hydroxide gel. The epitope lies within residues 19-37 of human alpha-CGRP1.
0.1 mL
Primary
796
Human, Rat