Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human Coagulation Panel 2 (3 plex) Non-distribution product as customer accommodation.
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Manufacturer:  Invitrogen™ EPX03010823901
Catalog No. 50-112-5799
Add to cart
Includes: Human Coagulation Panel 2 Reference Plasma Standard
Human Coagulaton Panel 2 Bead Mix
Human Coagulation Panel 2 Detection Antibody Mix
Reading Buffer
Wash Buffer 10x
Streptavidin-PE
Universal Assay Buffer 10X
Detection Antibody Diluent
PCR 8-Tube Strip
Adhesive Film
Greiner Microplate black 96 well
Description
Description
  • The human Coagulation panels are designed for the use with plasma samples (EDTA, Citrate)
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit does not contain a Multiscreen Filter Plate
Specifications
Specifications
Multiplex Immunoassay
Plasma (Citrat, EDTA)
Human
96 tests
Plasma (Citrat, EDTA): 25μL
Factor V, Factor VII, Factor VIII