Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human MMP-2 Simplex Non-distribution product as customer accommodation.
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Manufacturer:  Invitrogen™ EPX01A12132901
Catalog No. 50-112-5596
Add to cart
Includes: concentrated Magnetic Beads coated with monoclonal antibody to human MMP-2
human MMP-2 Standard (lyophilized)
Biotin-Conjugate (50x) anti-human MMP-2 monoclonal antibody
Universal Assay Buffer 10x
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • It is recommended to dilute MMP-2 samples when measured in serum or plasma
  • The recommended sample dilution factor is 1:50
Specifications
Specifications
Multiplex Immunoassay
Cell culture supernatant, serum, plasma
Human
96 tests
Serum, Plasma: 50μL (1:50 prediluted); CCS: 50μL
Matrix metalloproteinase-2