Learn More
Invitrogen™ eBioscience™ ProcartaPlex Mouse IFN alpha/IFN beta Panel (2 plex) Non-distribution product as customer accommodation.
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Manufacturer:  Invitrogen™ EPX02022187901
Catalog No. 50-112-5798
Add to cart
Includes: Mouse IFN alpha/IFN beta Standard Mix
Mouse IFN alpha/IFN beta Bead Mix
Human IFN alpha/IFN beta Detection Antibody Mix
Human IFN alpha/IFN beta Tertiary Antibody
Reading Buffer
Wash Buffer 10x
Streptavidin-PE
Universal Assay Buffer
Universal Assay Buffer 10x
Detection Antibody Diluent
PCR 8-Tube Strip
Adhesive Film
Greiner Microplate black 96 well
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit cannot be combined with other ProcartaPlex Simplex or Multiplex Kits
  • This kit does not contain a Multiscreen Filter Plate
Specifications
Specifications
Multiplex Immunoassay
Cell culture supernatant, serum, plasma
Mouse
96 tests
Serum, Plasma: 25μL; CCS: 50μL
IFN alpha, IFN beta