Learn More
Invitrogen™ eBioscience™ ProcartaPlex NHP IP-10 (CXCL10) Simplex Non-distribution product as customer accommodation.
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Manufacturer:  Invitrogen™ EPX01A40284901
Catalog No. 50-112-5743
Add to cart
Includes: concentrated Magnetic Beads coated with monoclonal antibody to non-human primate IP-10
non-human primate IP-10 Standard (lyophilized)
Biotin-Conjugate (50x) anti-non-human primate IP-10 monoclonal antibody
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
Specifications
Specifications
Multiplex Immunoassay
Cell culture supernatant, serum, plasma
Monkey
Interferon-gamma-inducible protein 10
96 tests
Serum, Plasma: 25μL; CCS: 50μL
CXCL10 (IP-10)