Learn More
GE Healthcare Processor Plus Gel Staining Tubing Kit Available on GSA/VA Contract for Federal Government customers only.
For use in gel processing.
Manufacturer:  GE Healthcare 80644556
Catalog No. 45-002-444
Add to cart
Description
Description
  • Kit includes PVC and tygon tubing, suction pipes, label, pump head dampeners
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
gel processing
10 x PVC Tubing 3.18mm I.D., 4.76mm O.D., 300mm L, 10 x Suction pipes, 10 x Label, marked 0-9, 1 x Tygon tubing 250mm L with barbed connector, coupling for inner pump head, 1 x Tygon tubing 250mm L with coupling, 1 x Tygon tubing 51mm L, 10 x Pump head dampeners
Gel Staining Tubing Kit