Learn More
Novus Biologicals™ Procine IL-18/IL-1F4 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP194192
Catalog No. NBP194192
Add to cart
Description
Description
The Porcine IL-18/IL-1F4 ELISA Kit (Colorimetric) measures porcine IL-18/IL-1F4 in cell culture supernatant, serum.
Specifications
Specifications
39.1 - 2,500pg/mL
3606
ELISA Kit
23.0pg/mL
Porcine (Po)
IL-18/IL-1F4
Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to pig IL-18, Biotin-Conjugate anti-pig IL-18 monoclonal antibody, Streptavidin-HRP, Pig IL-18 Standard lyophilized, 5ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
IL18