Learn More
DuPont ProClean 2 Blue Lab Coat, Traditional Collar
DuPont ProClean Apparel for Cleanrooms
Manufacturer: DuPont GB218SBUXL00300B
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
ProClean garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.
Specifications
Snaps | |
Knit | |
SMS | |
X-Large |
Blue | |
Controlled Environment | |
3 |