Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ ProClean™ 2 Shoe Cover, Case of 400
Click to view available options
Size:
2X-Large
Universal
X-Large
Description
ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.
Specifications
Size | Universal |
Material | SMS |
Color | Blue |
Sole Type | Non-skid |
For Use With (Application) | Controlled Environments |
Packaging Quantity | 400/Cs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction