Learn More
DuPont™ ProClean™ 2 Shoe Cover, Case of 500
DuPont™ ProClean™ Apparel for Cleanrooms
Supplier: DuPont™ GB440SBU0005000B
Description
ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.
Specifications
Universal | |
Blue | |
Controlled Environment |
SMS (Spunbound Melt-Blown Synthetic) | |
Heavy Duty Shoe Cover with Traction | |
500 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.