Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ 2 Shoe Cover, Case of 500

DuPont™ ProClean™ Apparel for Cleanrooms

Specifications

Material SMS (Spunbound Melt-Blown Synthetic)
Color Blue
Sole Type Heavy Duty Shoe Cover with Traction
For Use With (Application) Controlled Environment
Packaging Quantity 500
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-088-862
View Documents
DuPont™
GB440SBU0005000B
Universal This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-088-863
View Documents
DuPont™
GB440SBUXL05000B
X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-088-864
View Documents
DuPont™
GB440SBU2X05000B
2X-Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.

Specifications

Specifications

SMS (Spunbound Melt-Blown Synthetic)
Heavy Duty Shoe Cover with Traction
500
Blue
Controlled Environment
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit