Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProClean™ Microporous Composite Fabric Cleanroom Coveralls, Bulk Packed

Catalog Number p-3836502
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
5.11 to 6.3 ft.
5.5 to 5.10 ft.
5.8 to 6 ft.
6.1 to 6.5 ft.
6.2 to 6.6 ft.
6.3 to 7 ft.
Height (Metric):
155.75 to 192.04 cm
167.64 to 155.44 cm
176.78 to 182.88 cm
185.92 to 198.12 cm
188.97 to 201.16 cm
192.02 to 213.3 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Unique breathable composite material provides particulate and light splash protection for critical environments. Coveralls feature a zipper front, serged seams, and elastic wrists, ankles, and waist.

  • Lightweight
  • Anti-static treated
  • Low linting
  • Made from a microporous composite fabric
  • Serged seams
  • Bound neck
  • Elastic back
  • Covered elastic wrist and ankles
  • Zipper Closure

TRUSTED_SUSTAINABILITY
Product Type Cleanroom Coveralls
Size 3X-Large
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Back Style Elastic
Size (English) Chest 46 to 52 in.
Size (English) Inseam 32.5 in.
Size (Metric) Chest 116.8 to 132.08 cm
Size (Metric) Inseam 82.55 cm
Sleeve Style Set
Thickness 5.4 mil
For Use With (Application) Barrier to dry particles, microorganisms, and non-hazardous liquids in controlled environments
Length (English) Sleeve 40.5 in.
Length (Metric) Sleeve 102.87 cm
Height (English) 6.2 to 6.6 ft.
Height (Metric) 188.97 to 201.16 cm
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit