Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ ProCoat™ Cold Resistant PVC Gloves with Detachable Terry Liner
Foam insulation combined with a cold-resistant outer shell helps to keep hands warm and dry. A detachable, 7-gauge insulating liner provides the option for additional thermal protection.
$207.50 - $210.00
Specifications
Color | Blue |
---|---|
Exterior Finish | Fully Textured |
Glove Cuff Style | Open |
Length (English) | 12 in. |
Liner Material | Cotton |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-046-389
|
PIP™
588658DL/L |
Large |
Case of 72 for $207.50
|
|
19-046-390
|
PIP™
588658DL/XL |
X-Large |
Case of 72 for $207.50
|
|
19-046-391
|
PIP™
588658DL/XXL |
2X-Large |
Case of 72 for $210.00
|
|
Description
- Cold-resistant outer shell
- Foam insulation helps keep hands warm and dry
- Seamless knit construction protects hands without sacrificing comfort or dexterity
- Sandy finish enhances grip in wet and slippery conditions
- Detachable, insulating 7-gauge terry cloth liner
- Ideal for handling grease, oils, acids, caustics and solvents in cold environments
- Liquid-proof
Specifications
Blue | |
Open | |
Cotton | |
58-8658DL |
Fully Textured | |
12 in. | |
PVC | |
7 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title