Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRODH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554187
Description
Immunogen sequence: QESVAKLGIA SRAEIEDWFT AETLGVSGTM DLLDWSSLID SRTKLSKHLV VPNAQTGQLE PLLSRFTEEE ELQMTRMLQR MDVLAKKATE MGVRLMVDAE QTYFQ Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.
Specifications
PRODH | |
Polyclonal | |
Unconjugated | |
Prodh | |
HSPOX2; p53-induced gene 6 protein; PIG6; POX; POX2; Pro1; Pro-1; Prodh; PRODH 1; PRODH 2; PRODH1; PRODH2; proline dehydrogenase; proline dehydrogenase (oxidase) 1; proline dehydrogenase (proline oxidase); proline dehydrogenase 1; proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; SCZD4; TP53I6; tumor protein p53 inducible protein 6; Ym24d07 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5625 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43272 | |
Prodh | |
Recombinant protein corresponding to Human PRODH. Recombinant protein control fragment (Product #RP-92348). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction