Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human BMP-4 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$189.00 - $1649.00

Specifications

Gene Alias BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Source NS0
Conjugate Unlabeled
Recombinant Yes
Gene ID (Entrez) 652
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD31401 View Documents R&D Systems™
PRD31401
5μg Each for $189.00
Add to cart
 
PRD31410 View Documents R&D Systems™
PRD31410
50μg Each for $989.00
Add to cart
 
PRD31450 View Documents R&D Systems™
PRD31450
250μg Each for $1,649.00
Add to cart
 
Description

Description

The ProDots Recombinant Human BMP-4 Protein is derived from NS0. The ProDots Recombinant Human BMP-4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Unlabeled
652
Human
BMP4
<0.01 EU per 1μg of the protein by the LAL method.
Bioactivity
NS0
Yes
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Observed M.W.: 22-25kDa, reducing conditions 37-41, non-reducing conditions, Theoretical M.W.: 13kDa (monomer)
Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is typically 2.5-15.0ng/mL.
Q53XC5
Mouse myeloma cell line, NS0-derived Ser293 - Arg408
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit