Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human Flt-3 Ligand Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$759.00 - $1399.00

Specifications

Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Source Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived Thr27 - Pro185; N Terminal Sequence: Thr27
Conjugate Unlabeled
Recombinant Yes
Gene ID (Entrez) 2323
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD30810 View Documents R&D Systems™
PRD30810
50μg Each for $759.00
Add to cart
 
PRD30850 View Documents R&D Systems™
PRD30850
250μg Each for $1,399.00
Add to cart
 
Description

Description

The ProDots Recombinant Human Flt-3 Ligand Protein is derived from Sf 21 (baculovirus). The ProDots Recombinant Human Flt-3 Ligand Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Unlabeled
2323
Human
FLT3LG
<0.10 EU per 1μg of the protein by the LAL method.
Bioactivity
Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived Thr27 - Pro185; N Terminal Sequence: Thr27
Yes
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Predicted MW: 17.5kDa Observed MW: 17-30kDa, reducing conditions
Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is typically 0.2-1ng/mL.
AAA17999.1
Spodoptera frugiperda, Sf 21 (baculovirus)-derived Thr27 - Pro185
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit