Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ ProDots Human IFN-gamma Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer: R&D Systems™ PRD28550
This item is not returnable.
View return policy
Description
The ProDots Recombinant Human IFN-gamma Protein is derived from E. coli. The ProDots Recombinant Human IFN-gamma Protein has been validated for the following applications: Bioactivity.Specifications
CAA31639 | |
3458 | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain | |
E. coli-derived human IFN-gamma protein Gln24-Gln166 | |
<0.10 EU per 1μg of the protein by the LAL method. | |
IFN-gamma | |
Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is typically 0.15-0.75ng/mL. | |
Human | |
For a stock solution, reconstitute at 200μg/mL in sterile deionized water, or simply roll ProDot directly into cell culture medium for immediate use. |
Bioactivity | |
Predicted MW: 16.9kDa Observed MW: 17kDa, reducing conditions | |
250μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
IFNG | |
Unconjugated | |
Yes | |
E. coli-derived Gln24 - Gln166 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title