Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human IFN-gamma Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$179.00 - $409.00

Specifications

Gene Alias IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Source Source: E. coli-derived Gln24 - Gln166; N Terminal Sequence: Met
Conjugate Unlabeled
Recombinant Yes
Gene ID (Entrez) 3458
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD28510 View Documents R&D Systems™
PRD28510
50μg Each for $179.00
Add to cart
 
PRD28550 View Documents R&D Systems™
PRD28550
250μg Each for $409.00
Add to cart
 
Description

Description

The ProDots Recombinant Human IFN-gamma Protein is derived from E. coli. The ProDots Recombinant Human IFN-gamma Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Unlabeled
3458
Human
IFNG
<0.10 EU per 1μg of the protein by the LAL method.
Bioactivity
Source: E. coli-derived Gln24 - Gln166; N Terminal Sequence: Met
Yes
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Predicted MW: 16.9kDa Observed MW: 17kDa, reducing conditions
Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is typically 0.15-0.75ng/mL.
CAA31639
E. coli-derived Gln24 - Gln166
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit