Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human IL-10 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$299.00 - $1999.00

Specifications

Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Source Sf 21 (baculovirus)
Conjugate Unlabeled
Recombinant Yes
Gene ID (Entrez) 3586
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD21701 View Documents R&D Systems™
PRD21701
5μg Each for $299.00
Add to cart
 
PRD21710 View Documents R&D Systems™
PRD21710
50μg Each for $1,599.00
Add to cart
 
PRD21750 View Documents R&D Systems™
PRD21750
250μg Each for $1,999.00
Add to cart
 
Description

Description

The ProDots Recombinant Human IL-10 Protein is derived from Sf 21 (baculovirus). The ProDots Recombinant Human IL-10 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Unlabeled
3586
Human
IL10
<0.10 EU per 1μg of the protein by the LAL method.
Bioactivity
Sf 21 (baculovirus)
Yes
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Theoretical M.W.: 18.6kDa
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is typically 0.15-0.75ng/mL.
P22301
Spodoptera frugiperda, Sf 21 (baculovirus)-derived Ser19 - Asn178
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit