Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ ProDots Human IL-2 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Supplier: R&D Systems™ PRD20201
This item is not returnable.
View return policy
Description
The ProDots Recombinant Human IL-2 Protein is derived from E. coli. The ProDots Recombinant Human IL-2 Protein has been validated for the following applications: Bioactivity.Specifications
P60568 | |
3558 | |
5μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. | |
aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
IL2 | |
Unconjugated | |
Recombinant | |
E. coli-derived Ala21 - Thr153, with an N-terminal Met |
Bioactivity | |
Observed M.W.: 13kDa, reducing conditions, Theoretical M.W.: 15kDa | |
E. coli-derived human IL-2 protein Ala21-Thr153,with an N-terminal Met | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
IL-2 | |
Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295. The ED50 for this effect is typically 0.05-0.25ng/mL | |
Human | |
For a stock solution, reconstitute at 100 μg/mL in sterile 100 mM Acetic Acid, or simply roll ProDot™ directly into cell culture medium for immediate use. | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction