Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human IL-3 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$629.00 - $1099.00

Specifications

Gene Alias Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Source E. coli
Conjugate Unlabeled
Recombinant Yes
Gene ID (Entrez) 3562
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD20310 View Documents R&D Systems™
PRD20310
50μg Each for $629.00
Add to cart
 
PRD20350 View Documents R&D Systems™
PRD20350
250μg Each for $1,099.00
Add to cart
 
Description

Description

The ProDots Recombinant Human IL-3 Protein is derived from E. coli. The ProDots Recombinant Human IL-3 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Unlabeled
3562
Human
IL3
<0.10 EU per 1μg of the protein by the LAL method.
Bioactivity
E. coli
Yes
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Predicted MW: 15kDa Observed MW: 14kDa, reducing conditions
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is typically 0.02-0.1ng/mL.
Q6GS87
E. coli-derived Ala20 - Phe152, with and without an N-terminal Met
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit