Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Recombinant Human IL-6 Protein

Supplied as ready-to-use 5 ug protein balls

$857.65 - $1154.00

Specifications

Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Source E. coli-derived human IL-6 protein Pro29 - Met212
Conjugate Unconjugated
Recombinant Yes
Gene ID (Entrez) 3569
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD20610
View Documents
R&D Systems™
PRD20610
50 µg
Each for $857.65
Add to cart
 
PRD20650
View Documents
R&D Systems™
PRD20650
250 µg
Each for $1,154.00
Add to cart
 
Description

Description

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813.

The ED50 for this effect is 0.2-0.8 ng/mL.

Specifications

Specifications

B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Unconjugated
3569
Human
IL6
<0.10 EU / 1 µg of the protein by the LAL method.
Bioactivity
E. coli-derived human IL-6 protein Pro29 - Met212
Yes
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
M.W. (Observed): 21 kDa, reducing conditions; M.W. (theoretical): 20.3 kDa
Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is typically 0.2-0.8ng/mL.
Q75MH2
E. coli-derived Pro29 - Met212
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit