Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human LIF Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$629.00 - $1099.00

Specifications

Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Source Source: E. coli-derived Pro24 - Phe202; N Terminal Sequence: Pro24
Conjugate Unlabeled
Recombinant Yes
Gene ID (Entrez) 3976
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD773410 View Documents R&D Systems™
PRD773410
50μg Each for $629.00
Add to cart
 
PRD773450 View Documents R&D Systems™
PRD773450
250μg Each for $1,099.00
Add to cart
 
Description

Description

The ProDots Recombinant Human LIF Protein is derived from E. coli. The ProDots Recombinant Human LIF Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Unlabeled
3976
Observed M.W.: 115-137 kDa, reducing conditions; Theoretical M.W.: 19.6 kDa
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323.
P15018
E. coli-derived Pro24 - Phe202
Source: E. coli-derived Pro24 - Phe202; N Terminal Sequence: Pro24
Yes
Human
LIF
<0.01 EU per 1μg of the protein by the LAL method.
Bioactivity
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit