Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human PDGF-BB Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$169.00 - $1499.00

Specifications

Source E. coli
Conjugate Unlabeled
Recombinant Yes
Gene ID (Entrez) 5155
Purity >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD22001 View Documents R&D Systems™
PRD22001
5μg Each for $169.00
Add to cart
 
PRD22010 View Documents R&D Systems™
PRD22010
50μg Each for $849.00
Add to cart
 
PRD22050 View Documents R&D Systems™
PRD22050
250μg Each for $1,499.00
Add to cart
 
Description

Description

The ProDots Recombinant Human PDGF-BB Protein is derived from E. coli. The ProDots Recombinant Human PDGF-BB Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

E. coli
Yes
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Observed M.W.: 12kDa, reducing conditions, Theoretical M.W.: 12.3kDa (monomer)
Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is typically 1.5-6.0ng/mL.
Q6FHE7
E. coli-derived Ser82 - Thr190
Unlabeled
5155
Human
PDGFB
<0.10 EU per 1μg of the protein by the LAL method.
Bioactivity
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit