Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human VEGF 165 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$209.00 - $1154.00

Specifications

Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Conjugate Unconjugated
Recombinant Yes
Gene ID (Entrez) 7422
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRD29301
View Documents Promotion Details
R&D Systems™
PRD29301
5μg
Each for $209.00
Add to cart
 
PRD29310
View Documents Promotion Details
R&D Systems™
PRD29310
50μg
Each for $1,154.00
Add to cart
 
PRD29350
View Documents Promotion Details
R&D Systems™
PRD29350
250μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

The ProDots Recombinant Human VEGF 165 Protein is derived from Sf 21 (baculovirus). The ProDots Recombinant Human VEGF 165 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Unconjugated
7422
Human
VEGFA
<0.10 EU / 1 µg of the protein by the LAL method.
Bioactivity
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Yes
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Observed M.W.: 20-22kDa, reducing conditions 39-42kDa, non-reducing conditions, Theoretical M.W.: 19.2kDa (monomer)
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 is typically 1.0-6.0ng/mL.
NP_001165097
Spodoptera frugiperda, Sf 21 (baculovirus)-derived Ala27 - Arg191
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit