Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proDynorphin (GP Reactive) Guinea Pig anti-Guinea Pig, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$277.76 - $602.68

Specifications

Antigen proDynorphin (GP Reactive)
Concentration Conc. Not Determined
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0001 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP10109150UL
150 μL
Each for $602.68
Add to cart
 
50-210-0002 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1010950UL
50 μL
Each for $277.76
Add to cart
 
Description

Description

proDynorphin (GP Reactive) Polyclonal antibody specifically detects proDynorphin (GP Reactive) in Guinea Pig samples. It is validated for Immunohistochemistry
Specifications

Specifications

proDynorphin (GP Reactive)
Immunohistochemistry
Unconjugated
Guinea Pig
Guinea Pig
Q60478
100135596
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
ADCA; Alpha-neoendorphin; Beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); Dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; Leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; Proenkephalin-B; Rimorphin; SCA23
QEDPNAYSEEFFDV
Polyclonal
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Pdyn
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit