Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proDynorphin Guinea Pig anti-Mouse, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$283.96 - $602.68

Specifications

Antigen proDynorphin
Concentration 1 mg/mL
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0004 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1011050UL
50 μL
Each for $283.96
Add to cart
 
50-210-0003 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP10110150UL
150 μL
Each for $602.68
Add to cart
 
Description

Description

proDynorphin Polyclonal antibody specifically detects proDynorphin in Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

proDynorphin
Immunohistochemistry
Unconjugated
Guinea Pig
Mouse, Rat
O35417, P06300
18610, 29190
IgG
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Pdyn
1 mg/mL
Polyclonal
Liquid, Purified
RUO
PBS with 0.05% sodium azide
ADCA; Alpha-neoendorphin; Beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); Dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; Leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; Proenkephalin-B; Rimorphin; SCA23
SQENPNTYSEDLDV
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit