Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proenkephalin Mouse anti-All Species, Alexa Fluor™ 750, Clone: 1194/334, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB10062746AF750

Catalog No. N1062746A70

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Proenkephalin Monoclonal antibody specifically detects Proenkephalin in All Species samples. It is validated for ELISA.
Specifications

Specifications

Proenkephalin
1194/334
ELISA
enkephalin A, preproenkephalin, proenkephalin, proenkephalin-A
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This product recognises Met-enkephalin a neurotransmitter that both competes with, and mimics the effects of opiate drugs. Enkephalins play a role in a number of physiologic functions, including pain perception and responses to stress. Cross reactivates with other enkephalins to be determined.
ELISA
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
PENK
Native
0.1 mL
Primary
5179
All
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit